KEYH14DECD93 Key Organics / BIONET

Sulfobetaine

  • Cas Number : 4727-41-7
  • Molecular Weight : 120.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/862615

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-19261_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart