KEYH14DEDB7A Key Organics / BIONET

2-(2,6-Dioxopiperidin-3-yl)-4-nitroisoindoline-1,3-dione

  • Cas Number : 19171-18-7
  • Molecular Weight : 303.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/866175

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DS-5838_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart