KEYH14DEDC0E Key Organics / BIONET

(4-((Dimethylamino)methyl)phenyl)boronic acid hydrochloride

  • Cas Number : 938465-64-6
  • Molecular Weight : 215.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/866323

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DS-5986_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart