KEYH14DEE136 Key Organics / BIONET

4-Bromo-2-(hydroxymethyl)benzoic acid

  • Cas Number : 670256-21-0
  • Molecular Weight : 231.045

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/867643

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DS-7311_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart