KEYH14DEE54E Key Organics / BIONET

H-Orn(Boc)-OMe.HCl

  • MDL number MFCD00237207
  • Cas Number : 2480-96-8
  • Molecular Weight : 282.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 4
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/868691

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-8377_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart