KEYH14DEE62D Key Organics / BIONET

4-Chloro-2-phenylthieno[3,2-c]pyridine-7-carbonitrile

  • Cas Number : 1361197-82-1
  • Molecular Weight : 270.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/868914

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DS-8601_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart