KEYH14DEE90F Key Organics / BIONET

tert-Butyl (5-hydroxypyridin-2-yl)carbamate

  • MDL number MFCD09839686
  • Cas Number : 1187930-13-7
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 4
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/869649

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/DS-9339_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart