KEYH14DEECC2 Key Organics / BIONET

1-hydroxy-1-phenylpropan-2-one

  • Cas Number : 90-63-1
  • Molecular Weight : 150.177

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/870674

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NS-01150_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart