KEYH14DEFF3C Key Organics / BIONET

methyl 2-(2-nitrobenzylidene)acetoacetate

  • MDL number MFCD00551860
  • Cas Number : 39562-27-1
  • Molecular Weight : 249.222

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108698

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4438_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart