KEYH14DEFF3D Key Organics / BIONET

bis(2-oxa-6-azaspiro[3.3]heptan-6-ium) oxalate

  • MDL number MFCD16652332
  • Cas Number : 1045709-32-7
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-oxa-6-azaspiro[3.3]heptane ethanedioate (2:1)
Inchi Code 1S/2C5H9NO.C2H2O4/c2*1-5(2-6-1)3-7-4-5;3-1(4)2(5)6/h2*6H,1-4H2;(H,3,4)(H,5,6)
Inchi Key RXBYDYSXIHJXNO-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442215

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4866_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart