KEYH14DEFF3E Key Organics / BIONET

cubane-1,4-dicarboxylic acid

  • MDL number MFCD08458496
  • Cas Number : 32846-66-5
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,4-cubanedicarboxylic acid
Inchi Code 1S/C10H8O4/c11-7(12)9-1-2-4(9)6-5(9)3(1)10(2,6)8(13)14/h1-6H,(H,11,12)(H,13,14)
Inchi Key JFKXMJUMOJJTCQ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/870366

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-5129_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart