KEYH14DEFF58 Key Organics / BIONET

ethyl 4-amino-3,5-dibromobenzoate

  • MDL number MFCD00187798
  • Cas Number : 58922-06-8
  • Molecular Weight : 322.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1039512

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-5137_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart