KEYH14DEFF5A Key Organics / BIONET

ethyl 7-oxo-4,7-dihydropyrazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD00104600
  • Cas Number : 29274-18-8
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 7-hydroxypyrazolo[1,5-a]pyrimidine-6-carboxylate
Inchi Code 1S/C9H9N3O3/c1-2-15-9(14)6-5-10-7-3-4-11-12(7)8(6)13/h3-5,10H,2H2,1H3
Inchi Key KCRIHOOVSNHMLN-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1016469

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-32114_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart