KEYH2F621594 Key Organics / BIONET

5-Bromo-1-(difluoromethyl)pyridin-2(1H)-one

  • Cas Number : 832735-61-2
  • Molecular Weight : 224.005

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1046817

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FP-0056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart