KEYH2F621BFD Key Organics / BIONET

2-Boc-6-methoxy-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD13248619
  • Cas Number : 860436-57-3
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 6-methoxy-3,4-dihydroisoquinoline-2(1H)-carboxylate
Inchi Code 1S/C15H21NO3/c1-15(2,3)19-14(17)16-8-7-11-9-13(18-4)6-5-12(11)10-16/h5-6,9H,7-8,10H2,1-4H3
Inchi Key LQLAKQZVCKYEQC-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1043962

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-12678_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart