KEYH305DA1FF Key Organics / BIONET

4,4,5,5-tetramethyl-2-(3-propoxyphenyl)-1,3,2-dioxaborolane

  • Molecular Weight : 262.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1293973

Safety Information

signalword Warning
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AR-0086_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart