KEYH305DA20D Key Organics / BIONET

2-(6-methoxy-1-oxo-2,3-dihydro-1H-inden-2-yl)-2,3-dihydro-1H-isoindole-1,3-dione

  • Molecular Weight : 307.305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1095034

Safety Information

signalword Warning
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GP-0086_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart