KEYH305DA211 Key Organics / BIONET

tert-butyl N-(2-oxo-2,3-dihydro-1H-inden-5-yl)carbamate

  • Molecular Weight : 247.294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1095035

Safety Information

signalword Warning
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/HP-0010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart