KEYH305DA212 Key Organics / BIONET

2,2,2-trichloro-N-(2-methylbut-3-en-2-yl)acetamide

  • MDL number MFCD31742441
  • Cas Number : 91989-80-9
  • Molecular Weight : 230.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1095036

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/HP-0017_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart