KEYH305DA215 Key Organics / BIONET

3-chloro-N-hydroxy-2,2-dimethylpropanamide

  • Molecular Weight : 151.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1095038

Safety Information

signalword Warning
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KP-0067_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart