KEYH305DB57E Key Organics / BIONET

6-methoxy-5-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • Molecular Weight : 244.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store under N2, Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1294020

Safety Information

signalword Warning
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MP-0099_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart