KEYH305DB5B9 Key Organics / BIONET

6-methylcyclohex-1-ene-1-carboxylic acid

  • Molecular Weight : 140.182

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1095044

Safety Information

signalword Warning
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/MP-0032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart