KEYH305DB5C9 Key Organics / BIONET

1,4-dimethyl (2S,3S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxybutanedioate

  • MDL number MFCD31742595
  • Cas Number : 84107-20-0
  • Molecular Weight : 277.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1095049

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-5141_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart