KEYH326198D5 Key Organics / BIONET

2-(4-methylphenyl)malondialdehyde

  • MDL number MFCD00216506
  • Cas Number : 27956-35-0
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 135 - 137
Iupac Name (2E)-3-hydroxy-2-(4-methylphenyl)-2-propenal
Inchi Code 1S/C10H10O2/c1-8-2-4-9(5-3-8)10(6-11)7-12/h2-7,10H,1H3
Inchi Key TXQHUJPDCFREEC-UHFFFAOYSA-N

Description

Melting Point 135 - 137
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94947

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BB-0711_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart