KEYH57C55D46 Key Organics / BIONET

methyl 4-bromo-5-(difluoromethyl)thiophene-2-carboxylate

  • MDL number MFCD22200335
  • Cas Number : 1207557-47-8
  • Molecular Weight : 271.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-bromo-5-(difluoromethyl)-2-thiophenecarboxylate
Inchi Code 1S/C7H5BrF2O2S/c1-12-7(11)4-2-3(8)5(13-4)6(9)10/h2,6H,1H3
Inchi Key SYAUEMYKGPPNQA-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1357120

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/ER-0003_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart