KEYH57C55D4E Key Organics / BIONET

8-benzyl-2,8-diazaspiro[4.5]decane-1,4-dione

  • MDL number MFCD11043250
  • Cas Number : 1463-48-5
  • Molecular Weight : 258.321

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65710

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-1514_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart