KEYH57C55D5F Key Organics / BIONET

methyl 2-(2,4-dinitrophenyl)acetate

  • MDL number MFCD00047704
  • Cas Number : 58605-12-2
  • Molecular Weight : 240.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl (2,4-dinitrophenyl)acetate
Inchi Code 1S/C9H8N2O6/c1-17-9(12)4-6-2-3-7(10(13)14)5-8(6)11(15)16/h2-3,5H,4H2,1H3
Inchi Key HTANURMJHZJJSO-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1357129

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FH-0034_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart