KEYH57C560C0 Key Organics / BIONET

(4R,5R)-4,5-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-1,3-dioxolane

  • MDL number MFCD00138009
  • Cas Number : 3969-59-3
  • Molecular Weight : 302.367

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108609

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4330_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart