KEYH57C560EE Key Organics / BIONET

2-cyano-N-[3-(dimethylamino)propyl]acetamide

  • MDL number MFCD03392234
  • Cas Number : 15029-09-1
  • Molecular Weight : 169.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1357168

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-5241_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart