KEYH5DFE41AB Key Organics / BIONET

methyl 3-amino-5-chloro-1-benzothiophene-2-carboxylate

  • MDL number MFCD13689054
  • Cas Number : 1026241-99-5
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-amino-5-chlorobenzo[b]thiophene-2-carboxylate
Inchi Code 1S/C10H8ClNO2S/c1-14-10(13)9-8(12)6-4-5(11)2-3-7(6)15-9/h2-4H,12H2,1H3
Inchi Key CFYACBCHHBWDOO-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1435755

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/LR-0018_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart