KEYH93E4A792 Key Organics / BIONET

1,4-dioxaspiro[4.5]decan-8-yl (2S,3R)-2-(4-bromophenyl)-3-hydroxy-1-(4-methoxyphenyl)-3-methylpyrrolidine-2-carboxylate

  • MDL number MFCD30717725
  • Cas Number : 2307734-29-6
  • Molecular Weight : 546.458

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 4
Shipping Temperature Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665619

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/NS-00063_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart