KEYH93E4B5CF Key Organics / BIONET

ethyl 5-amino-1-[3-(trifluoromethyl)phenyl]-1H-pyrazole-4-carboxylate

  • Cas Number : 110821-29-9
  • Molecular Weight : 299.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1498248

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BT-0216_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart