LIF007177977 Life Chemicals Inc.

3-Acetyl-6-bromo-4-phenylquinolin-2(1H)-one

  • MDL number MFCD00186623
  • Cas Number : 139781-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart