LIF007177979 Life Chemicals Inc.

3-Acetyl-6-chloro-4-phenyl-1H-quinolin-2-one

  • MDL number MFCD00116018
  • Cas Number : 58375-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart