LIF007177992 Life Chemicals Inc.

2-(2-Oxo-2-phenyl-ethylsulfanyl)-benzoic acid

  • MDL number MFCD00425516
  • Cas Number : 25803-71-8
  • Molecular Weight : 272.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart