LIF007178012 Life Chemicals Inc.

2-hydrazinyl-6-methylpyrimidin-4(3H)-one

  • MDL number MFCD00193990
  • Cas Number : 37893-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart