LIF007178106 Life Chemicals Inc.

(2E)-3-[4-Methoxy-3-(piperidin-1-ylsulfonyl)phenyl]acrylic acid

  • MDL number MFCD00786080
  • Cas Number : 321979-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart