LIF007178143 Life Chemicals Inc.

Quinoxaline-5-carbaldehyde

  • MDL number MFCD00805833
  • Cas Number : 141234-08-4
  • Molecular Weight : 158.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart