LIF007178144 Life Chemicals Inc.

Quinoxaline-6-carbaldehyde

  • MDL number MFCD00805834
  • Cas Number : 130345-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart