LIF007178250 Life Chemicals Inc.

3-(3-Hydroxy-phenyl)-2-thioxo-thiazolidin-4-one

  • MDL number MFCD00022552
  • Cas Number : 23517-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart