LIF007178445 Life Chemicals Inc.

4-[(2-Chlorophenyl)amino]-4-oxobutanoic acid

  • MDL number MFCD00449646
  • Cas Number : 20266-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart