LIF007178446 Life Chemicals Inc.

N-(3-Trifluoromethyl-phenyl)-succinamic acid

  • MDL number MFCD00029839
  • Cas Number : 15386-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart