LIF007178448 Life Chemicals Inc.

N-(2,4-Dimethyl-phenyl)-succinamic acid

  • MDL number MFCD00029826
  • Cas Number : 76475-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart