LIF007178450 Life Chemicals Inc.

N-Pyridin-2-yl-succinamic acid

  • MDL number MFCD00030845
  • Cas Number : 62134-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart