LIF007178459 Life Chemicals Inc.

4-(Methylthio)-2-(4-oxo-2-thioxo-1,3-thiazolidin-3-yl)butanoic acid

  • MDL number MFCD00449880
  • Cas Number : 90812-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart