LIF007178607 Life Chemicals Inc.

(1-Isopropyl-1H-benzimidazol-2-yl)methanol

  • MDL number MFCD01942611
  • Cas Number : 305347-19-7
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart