LIF007178700 Life Chemicals Inc.

6-Bromo-naphthalen-2-ylamine

  • MDL number MFCD01026466
  • Cas Number : 7499-66-3
  • Molecular Weight : 222.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart