LIF007178865 Life Chemicals Inc.

1-(4-Methoxy-benzenesulfonyl)-piperidine-2-carboxylic acid

  • MDL number MFCD02656002
  • Cas Number : 253195-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart