LIF007178892 Life Chemicals Inc.

N-(4-Chloro-phenyl)-phthalamic acid

  • MDL number MFCD00029957
  • Cas Number : 7142-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart