LIF007178897 Life Chemicals Inc.

4-Amino-benzoic acid 2-oxo-2-phenyl-ethyl ester

  • MDL number MFCD00432455
  • Cas Number : 107692-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart